Merck
  • Production, purification, characterization, and applications of lipases.
Production, purification, characterization, and applications of lipases.
Biotechnology advances (2003-10-11)
R Sharma, Y Chisti, U C Banerjee
ABSTRACT
Lipases (triacylglycerol acylhydrolases, EC 3.1.1.3) catalyze the hydrolysis and the synthesis of esters formed from glycerol and long-chain fatty acids. Lipases occur widely in nature, but only microbial lipases are commercially significant. The many applications of lipases include speciality organic syntheses, hydrolysis of fats and oils, modification of fats, flavor enhancement in food processing, resolution of racemic mixtures, and chemical analyses. This article discusses the production, recovery, and use of microbial lipases. Issues of enzyme kinetics, thermostability, and bioactivity are addressed. Production of recombinant lipases is detailed. Immobilized preparations of lipases are discussed. In view of the increasing understanding of lipases and their many applications in high-value syntheses and as bulk enzymes, these enzymes are having an increasing impact on bioprocessing.
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)