Merck
Sex steroids and glucocorticoid ratios in Iberian lynx hair.
Conservation physiology (2020-08-28)
Alexandre Azevedo, Jella Wauters, Clemens Kirschbaum, Rodrigo Serra, António Rivas, Katarina Jewgenow
ABSTRACT
Knowledge on species' reproductive biology is a fundamental pre-requisite of every conservation effort, but is often lacking. Sex steroids can provide valuable information for the assessment of reproductive success, whereas glucocorticoids are used to assess adrenocortical activity and stress-related bodily adaption. However, due to their perilous condition, access to animals is often difficult, which makes hormone measurement in non-invasively collected hair samples an attractive option. We determined cortisol, cortisone, corticosterone, testosterone and progesterone in Iberian lynx hair using enzyme immunoassay (EIA). Cross-validation was performed with high-performance liquid chromatography (HPLC) and high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS). Finally, we statistically evaluated the variations of sex steroids and glucocorticoids according to age, sex, origin, behavior and management. All steroids except corticosterone were detectable in Iberian lynx hair. Hair progesterone measured by EIA was overestimated by cross-reaction with 5α-dihydroprogesterone, a biologically active gestagene, and was highly correlated with HPLC-MS/MS results. Progesterone was higher in adult females compared to all other age-sex groups. Cortisol measured by EIA was overestimated due to antibody cross-reactivity with cortisone and was correlated to the sum of HPLC-MS/MS measurements for cortisol and cortisone. Cortisol was higher in females than in males measured by HPLC-MS/MS, but the EIA results were confounded by the lack of specificity. When using cortisol-cortisone and cortisol-dihydroepiandrosterone ratios, differences were noted between wild-caught and captive-bred lynxes. Additionally, longitudinal EIA measurements of an Iberian lynx after a wildfire showed an inversion of the cortisol-cortisone ratio that later subsided. These results validate the use of hair progesterone measurement for Iberian lynx reproductive monitoring and add to the growing evidence supporting the need for a more comprehensive approach to hair steroid measurement that accounts for local interconversion and co-regulation mechanisms.
Sigma-Aldrich
Monoclonal Anti-Progesterone antibody produced in rat, clone 2H4, tissue culture supernatant