Merck
  • ECM deposition is driven by caveolin-1-dependent regulation of exosomal biogenesis and cargo sorting.
ECM deposition is driven by caveolin-1-dependent regulation of exosomal biogenesis and cargo sorting.
The Journal of cell biology (2020-10-15)
Lucas Albacete-Albacete, Inmaculada Navarro-Lérida, Juan Antonio López, Inés Martín-Padura, Alma M Astudillo, Alessia Ferrarini, Michael Van-Der-Heyden, Jesús Balsinde, Gertraud Orend, Jesús Vázquez, Miguel Ángel Del Pozo
ABSTRACT
The composition and physical properties of the extracellular matrix (ECM) critically influence tumor progression, but the molecular mechanisms underlying ECM layering are poorly understood. Tumor-stroma interaction critically depends on cell communication mediated by exosomes, small vesicles generated within multivesicular bodies (MVBs). We show that caveolin-1 (Cav1) centrally regulates exosome biogenesis and exosomal protein cargo sorting through the control of cholesterol content at the endosomal compartment/MVBs. Quantitative proteomics profiling revealed that Cav1 is required for exosomal sorting of ECM protein cargo subsets, including Tenascin-C (TnC), and for fibroblast-derived exosomes to efficiently deposit ECM and promote tumor invasion. Cav1-driven exosomal ECM deposition not only promotes local stromal remodeling but also the generation of distant ECM-enriched stromal niches in vivo. Cav1 acts as a cholesterol rheostat in MVBs, determining sorting of ECM components into specific exosome pools and thus ECM deposition. This supports a model by which Cav1 is a central regulatory hub for tumor-stroma interactions through a novel exosome-dependent ECM deposition mechanism.
Product Description
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vapa
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
U 18666A, powder
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Osbpl1a
Sigma-Aldrich
MISSION® esiRNA, targeting human CAV1
Sigma-Aldrich
Filipin III from Streptomyces filipinensis, ≥85% (HPLC)
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Tenascin antibody produced in rat, clone MTn-12, ascites fluid
Sigma-Aldrich
Anti-Tenascin Antibody, Chemicon®, from rabbit