Merck
  • FOXA2 controls the cis-regulatory networks of pancreatic cancer cells in a differentiation grade-specific manner.
FOXA2 controls the cis-regulatory networks of pancreatic cancer cells in a differentiation grade-specific manner.
The EMBO journal (2019-09-19)
Marta Milan, Chiara Balestrieri, Gabriele Alfarano, Sara Polletti, Elena Prosperini, Paola Spaggiari, Alessandro Zerbi, Giuseppe R Diaferia, Gioacchino Natoli
ABSTRACT
Differentiation of normal and tumor cells is controlled by regulatory networks enforced by lineage-determining transcription factors (TFs). Among them, TFs such as FOXA1/2 bind naïve chromatin and induce its accessibility, thus establishing new gene regulatory networks. Pancreatic ductal adenocarcinoma (PDAC) is characterized by the coexistence of well- and poorly differentiated cells at all stages of disease. How the transcriptional networks determining such massive cellular heterogeneity are established remains to be determined. We found that FOXA2, a TF controlling pancreas specification, broadly contributed to the cis-regulatory networks of PDACs. Despite being expressed in both well- and poorly differentiated PDAC cells, FOXA2 displayed extensively different genomic distributions and controlled distinct gene expression programs. Grade-specific functions of FOXA2 depended on its partnership with TFs whose expression varied depending on the differentiation grade. These data suggest that FOXA2 contributes to the regulatory networks of heterogeneous PDAC cells via interactions with alternative partner TFs.
Sigma-Aldrich
Anti-HNF1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-FOXA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution