Merck
  • Dysbindin-1 contributes to prefrontal cortical dendritic arbor pathology in schizophrenia.
Dysbindin-1 contributes to prefrontal cortical dendritic arbor pathology in schizophrenia.
Schizophrenia research (2018-05-16)
Glenn T Konopaske, Darrick T Balu, Kendall T Presti, Grace Chan, Francine M Benes, Joseph T Coyle
ABSTRACT
Deep layer III pyramidal cells in the dorsolateral prefrontal cortex (DLPFC) from subjects with schizophrenia and bipolar disorder previously were shown to exhibit dendritic arbor pathology. This study sought to determine whether MARCKS, its regulatory protein dysbindin-1, and two proteins, identified using microarray data, CDC42BPA and ARHGEF6, were associated with dendritic arbor pathology in the DLPFC from schizophrenia and bipolar disorder subjects. Using western blotting, relative protein expression was assessed in the DLPFC (BA 46) grey matter from subjects with schizophrenia (n = 19), bipolar disorder (n = 17) and unaffected control subjects (n = 19). Protein expression data were then correlated with dendritic parameter data obtained previously. MARCKS and dysbindin-1a expression levels did not differ among the three groups. Dysbindin-1b expression was 26% higher in schizophrenia subjects (p = 0.01) and correlated inversely with basilar dendrite length (r = -0.31, p = 0.048) and the number of spines per basilar dendrite (r = -0.31, p = 0.048), but not with dendritic spine density (r = -0.16, p = 0.32). The protein expression of CDC42BPA was 33% higher in schizophrenia subjects (p = 0.03) but, did not correlate with any dendritic parameter (p > 0.05). ARHGEF6 87 kDa isoform expression did not differ among the groups. CDC42BPA expression was not altered in frontal cortex from rats chronically administered haloperidol or clozapine. Dysbindin-1b appears to play a role in dendritic arbor pathology observed previously in the DLPFC in schizophrenia.
Sigma-Aldrich
Anti-DTNBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse