Merck
  • Rotenoids and flavonoids with anti-invasion of HT1080, anti-proliferation of U937, and differentiation-inducing activity in HL-60 from Erycibe expansa.
Rotenoids and flavonoids with anti-invasion of HT1080, anti-proliferation of U937, and differentiation-inducing activity in HL-60 from Erycibe expansa.
Bioorganic & medicinal chemistry (2006-12-13)
Hisashi Matsuda, Kazutoshi Yoshida, Katsutoshi Miyagawa, Yasunobu Asao, Saya Takayama, Souichi Nakashima, Fengming Xu, Masayuki Yoshikawa
ABSTRACT
Principal rotenoids (deguelin, tephrosin, rotenone, and 12a-hydroxyrotenone) (3-30microM) isolated from the stems of Erycibe expansa significantly inhibited invasion of human fibrosarcoma HT1080 cells through Matrigel-coated filters and release of proMMPs-2 and 9. In addition, deguelin and tephrosin showed differentiation-inducing activity in human promyelocytic leukemia HL-60 cells. Furthermore, effects of various constituents isolated from the ethyl acetate-soluble fraction on proliferation of human leukemia U937 cells were examined. As a result, most of isoflavones and several flavans as well as rotenoids showed moderate or substantial anti-proliferative activities.
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Tephrosin, ≥95% (LC/MS-ELSD)
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)