Merck
  • Intermedin protects thapsigargin‑induced endoplasmic reticulum stress in cardiomyocytes by modulating protein kinase A and sarco/endoplasmic reticulum Ca2+‑ATPase.
Intermedin protects thapsigargin‑induced endoplasmic reticulum stress in cardiomyocytes by modulating protein kinase A and sarco/endoplasmic reticulum Ca2+‑ATPase.
Molecular medicine reports (2020-12-11)
Zhidong Li, Jia Guo, Yunfei Bian, Mingsheng Zhang
ABSTRACT
Intermedin (IMD) is a calcitonin/calcitonin‑related peptide that elicits cardioprotective effects in a variety of heart diseases, such as cardiac hypertrophy and heart failure. However, the molecular mechanism of IMD remains unclear. The present study investigated the effects of IMD on neonatal rat ventricular myocytes treated with thapsigargin. The results of the present study demonstrated that thapsigargin induced apoptosis in cardiomyocytes in a dose‑ and time‑dependent manner. Thapsigargin induced endoplasmic reticulum stress, as determined by increased expression levels of 78‑kDa glucose‑regulated protein, C/EBP‑homologous protein and caspase‑12, which were dose‑dependently attenuated by pretreatment with IMD. In addition, IMD treatment counteracted the thapsigargin‑induced suppression of sarco/endoplasmic reticulum Ca2+‑ATPase (SERCA) activity and protein expression levels, and cytoplasmic Ca2+ overload. IMD treatment also augmented the phosphorylation of phospholamban, which is a crucial regulator of SERCA. Additionally, treatment with the protein kinase A antagonist H‑89 inhibited the IMD‑mediated cardioprotective effects, including SERCA activity restoration, anti‑Ca2+ overload, endoplasmic reticulum stress inhibition and antiapoptosis effects. In conclusion, the results of the present study suggested that IMD may protect cardiomyocytes against thapsigargin‑induced endoplasmic reticulum stress and the associated apoptosis at least partly by activating the protein kinase A/SERCA pathway.
Sigma-Aldrich
Monoclonal Anti-SERCa2 ATPase antibody produced in mouse, clone IID8
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Anti-PLN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Endoplasmic Reticulum Isolation Kit, isolation of intact ER from mammalian soft tissues and cultured cells
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film