Merck
  • Synthesis and evaluation of 3-aryloxymethyl-1,2-dimethylindole-4,7-diones as mechanism-based inhibitors of NAD(P)H:quinone oxidoreductase 1 (NQO1) activity.
Synthesis and evaluation of 3-aryloxymethyl-1,2-dimethylindole-4,7-diones as mechanism-based inhibitors of NAD(P)H:quinone oxidoreductase 1 (NQO1) activity.
Journal of medicinal chemistry (2007-10-20)
Marie A Colucci, Philip Reigan, David Siegel, Aurélie Chilloux, David Ross, Christopher J Moody
ABSTRACT
NAD(P)H:quinone oxidoreductase 1 is a proposed target in pancreatic cancer. We describe the synthesis of a series of indolequinones, based on the 5- and 6-methoxy-1,2-dimethylindole-4,7-dione chromophores with a range of phenolic leaving groups at the (indol-3-yl)methyl position. The ability of these indolequinones to function as mechanism-based inhibitors of purified recombinant human NQO1 was evaluated, as was their ability to inhibit both NQO1 and cell growth in human pancreatic MIA PaCa-2 tumor cells. The inhibition of rhNQO1 was related to the pKa of the leaving group: compounds with poorer phenolic leaving groups were poor inhibitors whereas those with more acidic leaving groups were more efficient inhibitors. These inhibition data also correlated with the inhibition NQO1 in MIA PaCa-2 cells. However, the data demonstrate that NQO1 inhibition does not correlate with growth inhibitory activity, at least in the MIA PaCa-2 cell line, suggesting that targets in addition to NQO1 need to be considered to explain the potent growth inhibitory activity of this series of indolequinones in human pancreatic cancer cells.
Sigma-Aldrich
4-Fluorophenol, 99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
4-Aminophenol, ≥98%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Hydroxypyridine, 97%
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
4-Cyanophenol, 95%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)