Merck
  • Synthesis, biochemical, and cellular evaluation of farnesyl monophosphate prodrugs as farnesyltransferase inhibitors.
Synthesis, biochemical, and cellular evaluation of farnesyl monophosphate prodrugs as farnesyltransferase inhibitors.
Journal of medicinal chemistry (2007-06-09)
Michelle K Clark, Sarah A Scott, Jonathan Wojtkowiak, Rosemarie Chirco, Patricia Mathieu, John J Reiners, Raymond R Mattingly, Richard F Borch, Richard A Gibbs
ABSTRACT
Certain farnesyl diphosphate (FPP) analogs are potent inhibitors of the potential anticancer drug target protein farnesyltransferase (FTase), but these compounds are not suitable as drug candidates. Thus, phosphoramidate prodrug derivatives of the monophosphate precursors of FPP-based FTase inhibitors have been synthesized. The monophosphates themselves were significantly more potent inhibitors of FTase than the corresponding FPP analogs. The effects of the prodrug 5b (a derivative of 3-allylfarnesyl monophosphate) have been evaluated on prenylation of RhoB and on the cell cycle in a human malignant schwannoma cell line (STS-26T). In combination treatments, 1-3 microM 5b plus 1 microM lovastatin induced a significant inhibition of RhoB prenylation, and a combination of these drugs at 1 microM each also resulted in significant cell cycle arrest in G1. Indeed, combinations as low as 50 nM lovastatin + 1 microM 5c or 250 nM lovastatin + 50 nM 5c were highly cytostatic in STS-26T cell culture.
Sigma-Aldrich
trans,trans-Farnesyl monophosphate ammonium salt, ≥95.0% (TLC)
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)