Merck
  • RNA editing enzyme APOBEC3A promotes pro-inflammatory M1 macrophage polarization.
RNA editing enzyme APOBEC3A promotes pro-inflammatory M1 macrophage polarization.
Communications biology (2021-01-24)
Emad Y Alqassim, Shraddha Sharma, A N M Nazmul H Khan, Tiffany R Emmons, Eduardo Cortes Gomez, Abdulrahman Alahmari, Kelly L Singel, Jaron Mark, Bruce A Davidson, A J Robert McGray, Qian Liu, Brian D Lichty, Kirsten B Moysich, Jianmin Wang, Kunle Odunsi, Brahm H Segal, Bora E Baysal
ABSTRACT
Pro-inflammatory M1 macrophage polarization is associated with microbicidal and antitumor responses. We recently described APOBEC3A-mediated cytosine-to-uracil (C > U) RNA editing during M1 polarization. However, the functional significance of this editing is unknown. Here we find that APOBEC3A-mediated cellular RNA editing can also be induced by influenza or Maraba virus infections in normal human macrophages, and by interferons in tumor-associated macrophages. Gene knockdown and RNA_Seq analyses show that APOBEC3A mediates C>U RNA editing of 209 exonic/UTR sites in 203 genes during M1 polarization. The highest level of nonsynonymous RNA editing alters a highly-conserved amino acid in THOC5, which encodes a nuclear mRNA export protein implicated in M-CSF-driven macrophage differentiation. Knockdown of APOBEC3A reduces IL6, IL23A and IL12B gene expression, CD86 surface protein expression, and TNF-α, IL-1β and IL-6 cytokine secretion, and increases glycolysis. These results show a key role of APOBEC3A cytidine deaminase in transcriptomic and functional polarization of M1 macrophages.
Sigma-Aldrich
MISSION® esiRNA, targeting human APOBEC3A
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution