Merck
  • Activation of dopamine receptor D1 inhibits glioblastoma tumorigenicity by regulating autophagic activity.
Activation of dopamine receptor D1 inhibits glioblastoma tumorigenicity by regulating autophagic activity.
Cellular oncology (Dordrecht) (2020-08-08)
Kang Yang, Minghai Wei, Zhaofei Yang, Zhenfa Fu, Ruixue Xu, Cheng Cheng, Xi Chen, Sheng Chen, Eric Dammer, Weidong Le
ABSTRACT
Recent studies have reported important roles of dopamine receptors in the early development and progression of glioblastoma (GBM). Here, we tested the antitumor activity of a Dopamine receptor D1 (DRD1) agonist, either alone or in combination with temozolomide (TMZ) on GBM cells. Immunofluorescence, immunohistochemistry and Western blotting were used to detect dopamine receptor expression in primary human GBM tissues. In addition, clinical data of GBM patients downloaded from The Cancer Genome Atlas (TCGA) were analyzed. Image-based tracking analysis of LC3 using a mCherry-eGFP-LC3 plasmid was utilized to monitor autophagic flux. Transmission electron microscopy (TEM) was used to visualize aggregation of autophagosomes/autolysosomes. Finally, DRD1 agonist (SKF83959)-induced inhibition of GBM growth was assessed in vitro and in vivo. Positive DRD1 expression was observed in human GBM tissues and found to be related with a good clinical outcome. DRD1 activation specifically inhibited GBM cell growth and significantly disrupted autophagic flux, which led to tumor cell death. Moreover, we found that DRD1 agonist treatment inhibited auto-lysosomal degradation in GBM cells and that this process was calcium overload dependent and related to inhibition of mammalian target of rapamycin (mTOR). Finally, we found that DRD1 agonist and TMZ co-treatment yielded a synergistic therapeutic effect both in vivo and in vitro. From our data we conclude that DRD1 activation inhibits GBM cell growth and may serve as an alternative avenue for the design of future GBM therapies.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
MISSION® esiRNA, targeting human DRD1
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)