Merck
N-Benzylbenzamides: a new class of potent tyrosinase inhibitors.
Bioorganic & medicinal chemistry letters (2006-03-04)
Sung Jin Cho, Jung Seop Roh, Won Suck Sun, Sung Han Kim, Ki Duk Park
ABSTRACT
A series of potent inhibitors of tyrosinase and their structure-activity relationships are described. N-Benzylbenzamide derivatives (1-21) with hydroxyl(s) were synthesized and tested for their tyrosinase inhibitory activity. With this series, compound 15 provided a potent tyrosinase inhibition: it effectively inhibited the oxidation of l-DOPA catalyzed by mushroom tyrosinase with an IC(50) of 2.2microM.
Sigma-Aldrich
N-Benzylbenzamide, ≥98%
Sigma-Aldrich
Kojic acid