Merck
  • LINC00675 activates androgen receptor axis signaling pathway to promote castration-resistant prostate cancer progression.
LINC00675 activates androgen receptor axis signaling pathway to promote castration-resistant prostate cancer progression.
Cell death & disease (2020-08-18)
Mengfei Yao, Xiaolei Shi, Yue Li, Yutian Xiao, William Butler, Yongqiang Huang, Leilei Du, Tianqi Wu, Xiaojie Bian, Guohai Shi, Dingwei Ye, Guohui Fu, Jianhua Wang, Shancheng Ren
ABSTRACT
The development of prostate cancer (PCa) from androgen-deprivation therapy (ADT) sensitive to castration resistant (CRPC) seriously impacts life quality and survival of PCa patients. Emerging evidence shows that long noncoding RNAs (lncRNAs) play vital roles in cancer initiation and progression. However, the inherited mechanisms of how lncRNAs participate in PCa progression and treatment resistance remain unclear. Here, we found that a long noncoding RNA LINC00675 was upregulated in androgen-insensitive PCa cell lines and CRPC patients, which promoted PCa progression both in vitro and in vivo. Knockdown of LINC00675 markedly suppressed tumor formation and attenuated enzalutamide resistance of PCa cells. Mechanistically, LINC00675 could directly modulate androgen receptor's (AR) interaction with mouse double minute-2 (MDM2) and block AR's ubiquitination by binding to it. Meanwhile, LINC00675 could bind to GATA2 mRNA and stabilize its expression level, in which GATA2 could act as a co-activator in the AR signaling pathway. Notably, we treated subcutaneous xenografts models with enzalutamide and antisense oligonucleotides (ASO) targeting LINC00675 in vivo and found that targeting LINC00675 would benefit androgen-deprivation-insensitive models. Our findings disclose that the LINC00675/MDM2/GATA2/AR signaling axis is a potential therapeutic target for CRPC patients.
Sigma-Aldrich
Adenine hemisulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
apo-Transferrin human, powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder