Merck
  • DNMT1-mediated methylation inhibits microRNA-214-3p and promotes hair follicle stem cell differentiate into adipogenic lineages.
DNMT1-mediated methylation inhibits microRNA-214-3p and promotes hair follicle stem cell differentiate into adipogenic lineages.
Stem cell research & therapy (2020-10-21)
Fangcao Jin, Min Li, Xuyang Li, Yunpeng Zheng, Kun Zhang, Xiaojun Liu, Bingjie Cai, Guangwen Yin
ABSTRACT
Dysfunction of the DNA methylation was associated with stem cell reprogramming. Moreover, DNA methyltransferase 1 (DNMT1) deficiency was involved in the differentiation of hair follicle stem cell (HFSc), but the molecular mechanisms remain unknown. HFSc from human scalp tissues were isolated and cultured. The oil red O staining was used to observe the adipogenesis. The interaction relationship between microRNA (miR)-214-3p and mitogen-activated protein kinase 1 (MAPK1) was accessed by dual-luciferase reporter gene assay. The methylation level of miR-214-3p promoter was detected by methylation-specific PCR and the enrichment of DNMT1 in miR-214-3p promoter by chromatin immunoprecipitation assay. A mouse model of trauma was established to observe the skin regeneration at 0, 6, and 14 days. Expression of DNMT1 and MAPK1 was increased in the HFSc, while the expression of miR-214-3p was reduced. Moreover, DNMT1 inhibited the expression of miR-214-3p by promoting the promoter methylation of miR-214-3p. Overexpression of DNMT1 could reduce the expression of miR-214-3p, but increase the expression of MAPK1 and the extent of extracellular signal regulated kinase (ERK)1/2 phosphorylation, leading to enhanced adipogenic differentiation. Importantly, DNMT1 promoted skin regeneration in vivo. Conversely, overexpression of miR-214-3p could reverse the effects of DNMT1 on adipogenesis of HFSc. DNMT1 promotes adipogenesis of HFSc by mediating miR-214-3p/MAPK1/p-ERK1/2 axis. This study may provide novel biomarkers for the potential application in stem cell therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT1