Merck
  • Expression of p11 and Heteromeric TASK Channels in Rat Carotid Body Glomus Cells and Nerve Growth Factor-differentiated PC12 Cells.
Expression of p11 and Heteromeric TASK Channels in Rat Carotid Body Glomus Cells and Nerve Growth Factor-differentiated PC12 Cells.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2020-09-05)
Hidetada Matsuoka, Mieczyslaw Pokorski, Keita Harada, Reiji Yoshimura, Masumi Inoue
ABSTRACT
TWIK-related acid-sensitive K+ (TASK) homomeric channels, TASK1 and TASK3, are present in PC12 cells. The channels do not heteromerize due plausibly to a lack of p11 protein. Single-channel recording reveals that most of the rat carotid body (CB) glomus cells express heteromeric TASK1-TASK3 channels, but the presence of p11 in glomus cells has not yet been verified. TASK1, but not TASK3, binds to p11, which has a retention signal for the endoplasmic reticulum. We hypothesized that p11 could facilitate heteromeric TASK1-TASK3 formation in glomus cells. We investigated this hypothesis in isolated immunocytochemically identified rat CB glomus cells. The findings were that glomus cells expressed p11-like immunoreactive (IR) material, and TASK1- and TASK3-like IR material mainly coincided in the cytoplasm. The proximity ligation assay showed that TASK1 and TASK3 heteromerized. In separate experiments, supporting evidence for the major role of p11 for channel heteromerization was provided in PC12 cells stimulated by nerve growth factor. p11 production took place there via multiple signaling pathways comprising mitogen-activated protein kinase and phospholipase C, and heteromeric TASK1-TASK3 channels were formed. We conclude that p11 is expressed and TASK1 and TASK3 heteromerize in rat CB glomus cells.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Nerve Growth Factor-2.5S from murine submaxillary gland, NGF-2.5S, lyophilized powder, suitable for cell culture