Merck
  • IgE Epitope Profiling for Allergy Diagnosis and Therapy - Parallel Analysis of a Multitude of Potential Linear Epitopes Using a High Throughput Screening Platform.
IgE Epitope Profiling for Allergy Diagnosis and Therapy - Parallel Analysis of a Multitude of Potential Linear Epitopes Using a High Throughput Screening Platform.
Frontiers in immunology (2020-10-30)
Thorsten Krause, Niels Röckendorf, Barbara Meckelein, Heike Sinnecker, Christian Schwager, Stefanie Möckel, Uta Jappe, Andreas Frey
ABSTRACT
Immunoglobulin E (IgE) is pivotal for manifestation and persistence of most immediate-type allergies and some asthma phenotypes. Consequently, IgE represents a crucial target for both, diagnostic purposes as well as therapeutic approaches. In fact, allergen-specific immunotherapy - aiming to re-route an IgE-based inflammatory response into an innocuous immune reaction against the allergen - is the only curative approach for IgE-mediated allergic diseases known so far. However, this requires the cognate allergen to be known. Unfortunately, even in well-characterized allergics or asthmatics, often just a small fraction of total IgE can be assigned to specific target allergens. To overcome this knowledge gap, we have devised an analytical platform for unbiased IgE target epitope detection. The system relies on chemically produced random peptide libraries immobilized on polystyrene beads ("one-bead-one-compound (OBOC) libraries") capable to present millions of different peptide motifs simultaneously to immunoglobulins from biological samples. Beads binding IgE are highlighted with a fluorophore-labeled anti-IgE antibody allowing fluorescence-based detection and isolation of positives, which then can be characterized by peptide sequencing. Setting-up this platform required an elaborate optimization process including proper choice of background suppressants, secondary antibody and fluorophore label as well as incubation conditions. For optimal performance our procedure involves a sophisticated pre-adsorption step to eliminate beads that react nonspecifically with anti-IgE secondary antibodies. This step turned out to be important for minimizing detection of "false positive" motifs that otherwise would erroneously be classified as IgE epitopes. In validation studies we were able to retrieve artificial test-peptide beads spiked into our library by using IgE directed against those test-peptides at physiological concentrations (≤20 IU/ml of specific IgE), and disease-relevant bead-bound epitopes of the major peanut allergen Ara h 2 by screening with sera from peanut allergics. Thus, we established a platform with which one can find and validate new immunoglobulin targets using patient material which displays a largely unknown immunoglobulin repertoire.
Sigma-Aldrich
Triisobutylsilane, 99%