Merck
  • Use of ZnAl-Layered Double Hydroxide (LDH) to Extend the Service Life of Reinforced Concrete.
Use of ZnAl-Layered Double Hydroxide (LDH) to Extend the Service Life of Reinforced Concrete.
Materials (Basel, Switzerland) (2020-04-15)
Celestino Gomes, Zahid Mir, Rui Sampaio, Alexandre Bastos, João Tedim, Frederico Maia, Cláudia Rocha, Mário Ferreira
ABSTRACT
This work investigated the use of ZnAl-layered double hydroxide (LDH) intercalated with nitrate or nitrite ions for controlling the corrosion of steel in reinforced concrete. The work started by analyzing the stability of the powder in the 1-14 pH range and the capacity for capturing chloride ions in aqueous solutions of different pH. The effect of the ZnAl-LDH on the corrosion of steel was studied in aqueous 0.05 M NaCl solution and in mortars immersed in 3.5% NaCl. It was found that the LDH powders dissolved partially at pH > 12. The LDH was able to capture chloride ions from the external solution, but the process was pH-dependent and stopped at high pH due to the partial dissolution of LDH and the preferential exchange of OH- ions. These results seemed to imply that ZnAl-LDH would not work in the alkaline environment inside the concrete. Nonetheless, preliminary results with mortars containing ZnAl-LDH showed lower penetration of chloride ions and higher corrosion resistance of the steel rebars.