Merck
Heterogeneously flagellated microswimmer behavior in viscous fluids.
Biomicrofluidics (2020-04-29)
Louis William Rogowski, Micah Oxner, Jiannan Tang, Min Jun Kim
ABSTRACT
An analysis of heterogeneously flagellated microswimmers inside viscous fluids is presented. Flagella harvested from Salmonella typhimurium were isolated, repolymerized, and functionalized to have biotin at their ends, allowing for chemical attachment along the surfaces of avidin-coated microparticles. Assembled microswimmers were rotated under incremental magnetic field frequencies, in saline and methylcellulose solutions, to baseline their velocity responses. A mean square displacement analysis revealed that rotating microswimmers exhibited anomalous diffusive behavior at small time scales in each fluid and had increased diffusivity compared with the non-rotating cases. Flagellated microswimmers had decreased diffusivity when compared with non-flagellated microparticles in Brownian conditions. Microswimmers were demonstrated to perform selected trajectories under proportional feedback control with reasonable accuracy. Finally, microswimmer propulsion was shown to be heavily influenced by the handedness of the rotating magnetic fields, with frequency induced reversals of swimming direction observed under clockwise rotation; this effect was determined to be the result of flagellar bundling and unbundling.
Sigma-Aldrich
Cy®3 Mono 5-pack, Cytiva PA23001
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%