Merck
  • TRPA1 activation mediates nociception behaviors in a mouse model of relapsing-remitting experimental autoimmune encephalomyelitis.
TRPA1 activation mediates nociception behaviors in a mouse model of relapsing-remitting experimental autoimmune encephalomyelitis.
Experimental neurology (2020-02-12)
Diéssica Padilha Dalenogare, Maria Carolina Theisen, Diulle Spat Peres, Maria Fernanda Pessano Fialho, Débora Denardin Lückemeyer, Caren Tatiane de David Antoniazzi, Sabrina Qader Kudsi, Marcella de Amorim Ferreira, Camila Dos Santos Ritter, Juliano Ferreira, Sara Marchesan Oliveira, Gabriela Trevisan
ABSTRACT
Central neuropathic pain is the main symptom caused by spinal cord lesion in relapsing-remitting multiple sclerosis (RRMS), but its management is still not effective. The transient receptor potential ankyrin 1 (TRPA1) is a pain detecting ion channel involved in neuropathic pain development. Thus, the aim of our study was to evaluate the role of TRPA1 in central neuropathic nociception induced by relapsing-remitting experimental autoimmune encephalomyelitis (RR-EAE) mouse model. In this model, we observed the development of similar clinical conditions of RRMS in C57BL/6 female mice through RR-EAE using MOG35-55 antigen and Quil A adjuvant. At the thirty-fifth day post-induction, C57BL/6 female mice demonstrated alteration in the RR-EAE score without motor impairment, mechanical and cold allodynia. Also, significative changes in demyelinating (Mog and olig-1) and neuroinflammatory (Iba1, Gfap and Tnfa) markers were observed, but this model did not alter Trpa1 RNA expression levels in the spinal cord. The hydrogen peroxide and 4-hydroxynonenal levels (TRPA1 agonists) were increased in RR-EAE induced mice, as well as the NADPH oxidase activity. The intragastric treatment of RR-EAE induced mice with TRPA1 antagonists (HC-030031 and A-967079) and antioxidant (α-lipoic acid and apocynin) caused an antiallodynic effect. Moreover, the intrathecal administration of TRPA1 antisense oligonucleotide, HC-030031, α-lipoic acid, and apocynin transiently attenuated mechanical and cold allodynia. Thus, TRPA1 plays a key role in the induction of neuropathic pain in this model of RR-EAE and can be a possible target for investigating the development of pain in RRMS patients.
Sigma-Aldrich
Cytochrome c Reductase (NADPH) Assay Kit, 1 kit sufficient for 100 tests, determining cytochrome c reductase activity
Pricing and availability is not currently available.