Merck
  • CTCF is dispensable for immune cell transdifferentiation but facilitates an acute inflammatory response.
CTCF is dispensable for immune cell transdifferentiation but facilitates an acute inflammatory response.
Nature genetics (2020-06-10)
Grégoire Stik, Enrique Vidal, Mercedes Barrero, Sergi Cuartero, Maria Vila-Casadesús, Julen Mendieta-Esteban, Tian V Tian, Jinmi Choi, Clara Berenguer, Amaya Abad, Beatrice Borsari, François le Dily, Patrick Cramer, Marc A Marti-Renom, Ralph Stadhouders, Thomas Graf
ABSTRACT
Three-dimensional organization of the genome is important for transcriptional regulation1-7. In mammals, CTCF and the cohesin complex create submegabase structures with elevated internal chromatin contact frequencies, called topologically associating domains (TADs)8-12. Although TADs can contribute to transcriptional regulation, ablation of TAD organization by disrupting CTCF or the cohesin complex causes modest gene expression changes13-16. In contrast, CTCF is required for cell cycle regulation17, embryonic development and formation of various adult cell types18. To uncouple the role of CTCF in cell-state transitions and cell proliferation, we studied the effect of CTCF depletion during the conversion of human leukemic B cells into macrophages with minimal cell division. CTCF depletion disrupts TAD organization but not cell transdifferentiation. In contrast, CTCF depletion in induced macrophages impairs the full-blown upregulation of inflammatory genes after exposure to endotoxin. Our results demonstrate that CTCF-dependent genome topology is not strictly required for a functional cell-fate conversion but facilitates a rapid and efficient response to an external stimulus.
Product Description
ECL Start Western Blotting Detection Reagent, Cytiva RPN3243, pkg of 200 mL
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.