Merck
  • Reference values for salivary cortisol in healthy young infants by liquid chromatography-tandem mass spectrometry.
Reference values for salivary cortisol in healthy young infants by liquid chromatography-tandem mass spectrometry.
Pediatrics international : official journal of the Japan Pediatric Society (2020-01-25)
Mayako Saito-Abe, Kiwako Yamamoto-Hanada, Shoji F Nakayama, Yuki Hashimoto, Osamu Natsume, Maki Fukami, Tomonobu Hasegawa, Yukihiro Ohya
ABSTRACT
Sampling of salivary cortisol is non-invasive and important for the evaluation of the hypothalamic-pituitary-adrenal axis function and stress levels. However, the reference values for salivary cortisol measured by liquid chromatography-tandem mass spectrometry (LC-MS/MS) in healthy infants are unclear. The aim of this study was to establish the reference values for salivary cortisol levels in healthy infants. This study was a prospective observational cohort study following the participants until the age of 6 months. We analyzed 71 healthy, full-term infants at age 1 month between December 2017 and March 2018. We repeated saliva sampling every month, measured the salivary cortisol levels in the early morning by LC-MS/MS, and took the subjects' medical history by questionnaire. The minimum, 25th, 50th, 75th percentile, and maximum salivary cortisol levels were 0.08, 1.11, 2.21, 5.18, and 30.35 nmol/L, respectively. We established the reference values for salivary cortisol in young infants using LC-MS/MS.
Sigma-Aldrich
Cortisol-9,11,12,12-d4, ≥98 atom % D, ≥98% (CP)