Merck
  • Benzoselenadiazole-Based Conjugated Molecules: Active Switching Layers with Nanofibrous Morphology for Nonvolatile Organic Resistive Memory Devices.
Benzoselenadiazole-Based Conjugated Molecules: Active Switching Layers with Nanofibrous Morphology for Nonvolatile Organic Resistive Memory Devices.
ChemPlusChem (2020-05-14)
Rohit G Jadhav, Amitesh Kumar, Sanjay Kumar, Sayan Maiti, Shaibal Mukherjee, Apurba K Das
ABSTRACT
In this work, two symmetrical donor-acceptor-donor (D-A-D) type benzoselenadiazole (BSeD)-based π-conjugated molecules were synthesized and employed as an active switching layer for non-volatile data storage applications. BSeD-based derivatives with different donor units attached through common vinylene linkers showed different electrical and optical properties. 4,7-Di((E)-styryl)benzo[c][2,1,3]selenadiazole (DSBSeD) and 4,7-bis((E)-4-methoxystyryl)benzo[c][2,1,3]selenadiazole (DMBSeD) are sandwiched between gallium-doped ZnO (GZO) and metal aluminum electrodes respectively through solution-processed spin-coating method. The solution-processed nanofibrous switching layer containing the DMBSeD-based memory device showed reliable memory characteristics in terms of write and erase operations with low SET voltage than the random-aggregated DSBSeD-based device. The nanofibrous molecular morphology of switching layer overcomes the interfacial hole transport energy barrier at the interface of the DMBSeD thin-film and the bottom GZO electrode. The memory device GZO/DMBSeD/Al based on nanofibrous switching layers shows switching characteristics at compliance current of 10 mA with Vset =0.79 V and Vreset =-0.55 V. This work will be beneficial for the rational design of advanced next-generation organic memory devices by controlling the nanostructured morphology of active organic switching layer for enhanced charge-transfer phenomenon.
Sigma-Aldrich
4-Vinylanisole, 97%