Merck
  • Small Molecule Enhancers of Endosome-to-Cytosol Import Augment Anti-tumor Immunity.
Small Molecule Enhancers of Endosome-to-Cytosol Import Augment Anti-tumor Immunity.
Cell reports (2020-07-16)
Patrycja Kozik, Marine Gros, Daniel N Itzhak, Leonel Joannas, Sandrine Heurtebise-Chrétien, Patrycja A Krawczyk, Pablo Rodríguez-Silvestre, Andrés Alloatti, Joao Gamelas Magalhaes, Elaine Del Nery, Georg H H Borner, Sebastian Amigorena
ABSTRACT
Cross-presentation of antigens by dendritic cells (DCs) is critical for initiation of anti-tumor immune responses. Yet, key steps involved in trafficking of antigens taken up by DCs remain incompletely understood. Here, we screen 700 US Food and Drug Administration (FDA)-approved drugs and identify 37 enhancers of antigen import from endolysosomes into the cytosol. To reveal their mechanism of action, we generate proteomic organellar maps of control and drug-treated DCs (focusing on two compounds, prazosin and tamoxifen). By combining organellar mapping, quantitative proteomics, and microscopy, we conclude that import enhancers undergo lysosomal trapping leading to membrane permeation and antigen release. Enhancing antigen import facilitates cross-presentation of soluble and cell-associated antigens. Systemic administration of prazosin leads to reduced growth of MC38 tumors and to a synergistic effect with checkpoint immunotherapy in a melanoma model. Thus, inefficient antigen import into the cytosol limits antigen cross-presentation, restraining the potency of anti-tumor immune responses and efficacy of checkpoint blockers.
Sigma-Aldrich
Eeyarestatin I, ≥98% (HPLC)
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
DBeQ, ≥98% (HPLC)
Sigma-Aldrich
Importazole, ≥98% (HPLC)
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
E-64, protease inhibitor