Merck
Crack path selection in orientationally ordered composites.
Physical review. E (2020-08-17)
A Mesgarnejad, C Pan, R M Erb, S J Shefelbine, A Karma
ABSTRACT
While cracks in isotropic homogeneous materials propagate straight, perpendicularly to the tensile axis, cracks in natural and synthetic composites deflect from a straight path, often increasing the toughness of the material. Here we combine experiments and simulations to identify materials properties that predict whether cracks propagate straight or kink on a macroscale larger than the composite microstructure. Those properties include the anisotropy of the fracture energy, which we vary several fold by increasing the volume fraction of orientationally ordered alumina (Al_{2}O_{3}) platelets inside a polymer matrix, and a microstructure-dependent process zone size that is found to modulate the additional stabilizing or destabilizing effect of the nonsingular stress acting parallel to the crack. Those properties predict the existence of an anisotropy threshold for crack kinking and explain the surprisingly strong dependence of this threshold on sample geometry and load distribution.
Sigma-Aldrich
Isobornyl acrylate, technical grade, contains 200 ppm monomethyl ether hydroquinone as inhibitor