Merck
  • PDE1B2 regulates cGMP and a subset of the phenotypic characteristics acquired upon macrophage differentiation from a monocyte.
PDE1B2 regulates cGMP and a subset of the phenotypic characteristics acquired upon macrophage differentiation from a monocyte.
Proceedings of the National Academy of Sciences of the United States of America (2006-01-13)
Andrew T Bender, Joseph A Beavo
ABSTRACT
Monocyte-to-macrophage differentiation with the cytokine granulocyte-macrophage colony-stimulating factor induces expression of the cyclic nucleotide phosphodiesterase PDE1B2. However, what role PDE1B2 plays in macrophage biology has not been elucidated. We have addressed this question by inhibiting PDE1B2 induction by using RNA interference. Using a retrovirus-based system, we created HL-60 stable cell lines that express a short-hairpin RNA targeting PDE1B2. HL-60 cells treated with phorbol-12-myristate-13-acetate differentiate to a macrophage-like phenotype and up-regulate PDE1B2. However, expression of PDE1B2 short hairpin RNA effectively suppresses PDE1B2 mRNA, protein, and activity up-regulation. Using the HL-60 PDE1B2 knockdown cells and agonists for either adenylyl or guanylyl cyclase, it was found that PDE1B2 predominantly regulates cGMP and plays a lesser role in cAMP regulation in response to cyclase agonists. Furthermore, in intact HL-60 cells, PDE1B2 activity can be regulated by changes in Ca+2 levels. Inhibiting PDE1B2 up-regulation does not prevent HL-60 cell differentiation, because several markers of macrophage differentiation are unaffected. However, suppression of PDE1B2 expression alters some aspects of the macrophage-like phenotype, because cell spreading, phagocytic ability, and CD11b expression are augmented. The cAMP analog 8-Bromo-cAMP reverses the changes caused by PDE1B2 knockdown. Also, PDE1B2 knockdown cells have lower basal levels of cAMP and alterations in the phosphorylation state of several probable PKA substrate proteins. Thus, the effects of PDE1B2 on differentiation may ultimately be mediated through decreased cAMP. In conclusion, PDE1B2 regulates a subset of phenotypic changes that occur upon phorbol-12-myristate-13-acetate-induced differentiation and likely also plays a role in differentiated macrophages by regulating agonist-stimulated cGMP levels.
Sigma-Aldrich
PDE1B active rat, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)