- Non-invasive detection of candidate pregnancy protein biomarkers in the feces of captive polar bears (Ursus maritimus).
Non-invasive detection of candidate pregnancy protein biomarkers in the feces of captive polar bears (Ursus maritimus).
Currently, there is no method of accurately and non-invasively diagnosing pregnancy in polar bears. Specific proteins may exhibit altered profiles in the feces of pregnant bears, but predicting appropriate candidate proteins to investigate is speculative at best. The objective of this study was to identify potential pregnancy biomarker proteins based on their increased abundance in the feces of pregnant polar bears compared to pseudopregnant females (controls) using two-dimensional in-gel electrophoresis (2D-DIGE) and mass spectrometry (MS). Three 2D-DIGE gels were performed to evaluate fecal protein profiles from controls (n=3) and pregnant polar bears (n=3). There were 2224.67±52.39 (mean±SEM) spots resolved per gel. Of these, only five proteins were elevated in the pregnant group (P<0.05), and seven additional spots tended to be higher (0.05<P<0.10). All 12 were submitted for MS analysis and the identities of 11 were ascertained with a >99.9% confidence interval. The 11 spots represented seven distinct proteins, five of which were significantly more abundant in the pregnant group: IgGFc-binding protein, filamin-C, carboxypeptidase B, transthyretin, and immunoglobulin heavy chain variable region. To our knowledge, this was the first study that employed 2D-DIGE to identify differentially expressed proteins in fecal samples to characterize a physiological condition other than those related to gastrointestinal disorders. These promising results provided a strong foundation for ensuing efforts to develop a non-invasive pregnancy assay for use in both captive and wild polar bears.