Merck
  • Puromycin resistance gene as an effective selection marker for ciliate Tetrahymena.
Puromycin resistance gene as an effective selection marker for ciliate Tetrahymena.
Gene (2013-11-05)
Masaaki Iwamoto, Chie Mori, Yasushi Hiraoka, Tokuko Haraguchi
ABSTRACT
A puromycin-N-acetyltransferase gene (pac) is widely used as a selection marker for eukaryotic gene manipulation. However, it has never been utilized for molecular studies in the ciliate Tetrahymena thermophila, in spite of the limited number of selection markers available for this organism. To utilize pac as a maker gene for T. thermophila, the nucleotide sequence of the pac gene was altered to accord with the most preferred codon-usage in T. thermophila. This codon-optimized pac gene expressed in T. thermophila conferred a resistance to transformed cells against 2,000 μg/ml of puromycin dihydrochloride, whereas the growth of wild-type cells was completely inhibited by 200 μg/ml. Furthermore, an expression cassette constructed with the codon-optimized pac and an MTT1 promoter was effectively utilized for experiments to tag endogenous proteins of interest by fusing the cassette into the target gene locus. These results indicate that pac can be used as a selection marker in molecular studies of T. thermophila.
Sigma-Aldrich
Puromycin ready made solution, 10 mg/mL in H2O, ≥98% (HPLC), Cell culture tested