Merck
  • Liver-Targeted Small-Molecule Inhibitors of Proprotein Convertase Subtilisin/Kexin Type 9 Synthesis.
Liver-Targeted Small-Molecule Inhibitors of Proprotein Convertase Subtilisin/Kexin Type 9 Synthesis.
Angewandte Chemie (International ed. in English) (2017-10-27)
Kim F McClure, David W Piotrowski, Donna Petersen, Liuqing Wei, Jun Xiao, Allyn T Londregan, Adam S Kamlet, Anne-Marie Dechert-Schmitt, Brian Raymer, Roger B Ruggeri, Daniel Canterbury, Chris Limberakis, Spiros Liras, Paul DaSilva-Jardine, Robert G Dullea, Paula M Loria, Benjamin Reidich, Christopher T Salatto, Heather Eng, Emi Kimoto, Karen Atkinson, Amanda King-Ahmad, Dennis Scott, Kevin Beaumont, Jeffrey R Chabot, Michael W Bolt, Kevin Maresca, Kenneth Dahl, Ryosuke Arakawa, Akihiro Takano, Christer Halldin
ABSTRACT
Targeting of the human ribosome is an unprecedented therapeutic modality with a genome-wide selectivity challenge. A liver-targeted drug candidate is described that inhibits ribosomal synthesis of PCSK9, a lipid regulator considered undruggable by small molecules. Key to the concept was the identification of pharmacologically active zwitterions designed to be retained in the liver. Oral delivery of the poorly permeable zwitterions was achieved by prodrugs susceptible to cleavage by carboxylesterase 1. The synthesis of select tetrazole prodrugs was crucial. A cell-free in vitro translation assay containing human cell lysate and purified target mRNA fused to a reporter was used to identify active zwitterions. In vivo PCSK9 lowering by oral dosing of the candidate prodrug and quantification of the drug fraction delivered to the liver utilizing an oral positron emission tomography 18 F-isotopologue validated our liver-targeting approach.
Sigma-Aldrich
PF-06815345, ≥98% (HPLC)