Merck
  • A new congenital multicore titinopathy associated with fast myosin heavy chain deficiency.
A new congenital multicore titinopathy associated with fast myosin heavy chain deficiency.
Annals of clinical and translational neurology (2020-04-21)
Aurélien Perrin, Corinne Metay, Marcello Villanova, Robert-Yves Carlier, Elena Pegoraro, Raul Juntas Morales, Tanya Stojkovic, Isabelle Richard, Pascale Richard, Norma B Romero, Henk Granzier, Michel Koenig, Edoardo Malfatti, Mireille Cossée
ABSTRACT
Congenital titinopathies are myopathies with variable phenotypes and inheritance modes. Here, we fully characterized, using an integrated approach (deep phenotyping, muscle morphology, mRNA and protein evaluation in muscle biopsies), two siblings with congenital multicore myopathy harboring three TTN variants predicted to affect titin stability and titin-myosin interactions. Muscle biopsies showed multicores, type 1 fiber uniformity and sarcomeric structure disruption with some thick filament loss. Immunohistochemistry and Western blotting revealed a marked reduction of fast myosin heavy chain isoforms. This is the first observation of a titinopathy suggesting that titin defect leads to secondary loss of fast myosin heavy chain isoforms.
Sigma-Aldrich
Monoclonal Anti-TTN antibody produced in mouse, clone 7D3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid