Merck
  • Enantioselective analysis of secondary alcohols and their esters in purple and yellow passion fruits.
Enantioselective analysis of secondary alcohols and their esters in purple and yellow passion fruits.
Journal of agricultural and food chemistry (2007-11-17)
Hedwig Strohalm, Márta Dregus, Astrid Wahl, Karl-Heinz Engel
ABSTRACT
The enantiomeric compositions of the acetates, butanoates, hexanoates, and octanoates of the secondary alcohols 2-pentanol, 2-heptanol, and 2-nonanol were determined in yellow (Passiflora edulis f. flavicarpa) and purple (Passiflora edulis Sims) passion fruits. The compounds were isolated by means of simultaneous distillation-extraction. Enantiodifferentiation was performed via multidimensional gas chromatography using heptakis(2,3-di-O-methyl-6-O-tert-butyldimethylsilyl)-beta-cyclodextrin as chiral stationary phase. The series of homologous 2-alkyl esters, which are typical constituents of purple passion fruits, were shown to be present as nearly optically pure (R)-enantiomers. The proportions of the (S)-enantiomers varied in different batches and were dependent on the alcohol moieties of the esters. For minor amounts of esters detected in yellow fruits, the (R)-enantiomers were also dominating. However, the enantiomeric excesses were significantly lower than in the purple variety. Enantioselective analysis of the free alcohols revealed that 2-heptanol exhibited opposite configurations in purple and yellow passion fruits. A similar phenomenon was observed for 2-pentanol, which was present in the yellow fruits as a nearly racemic mixture. Data determined in extracts obtained by other techniques (liquid-liquid extraction, vacuum headspace technique) showed that the isolation procedure had no significant impact on the enantiomeric ratios.