Merck
  • Di-(2-ethylhexyl) phthalate inhibits expression and internalization of transthyretin in human placental trophoblastic cells.
Di-(2-ethylhexyl) phthalate inhibits expression and internalization of transthyretin in human placental trophoblastic cells.
Toxicology and applied pharmacology (2020-03-24)
Zhi-Ping Du, Shun Feng, Yan-Ling Li, Rong Li, Jia Lv, Wen-Qiang Ren, Qiang-Wei Feng, Ping Liu, Qu-Nan Wang
ABSTRACT
During pregnancy, fetal thyroid hormones (THs) are dependent on maternal placental transport and their physiological level is crucial for normal fetal neurodevelopment. Earlier research has shown that Di-(2-ethylhexyl) phthalate (DEHP) disrupts thyroid function and THs homeostasis in pregnant women and fetuses, and affects placental THs transport. However, the underlying mechanisms are poorly understood. The present study, therefore, aimed to systematically investigate the potential mechanisms of DEHP-induced disruption in the placental THs transport using two human placental trophoblastic cells, HTR-8/SVneo cells and JEG-3 cells. While the exposure of DEHP at the doses of 0-400 μM for 24 h did not affect cell viability, we found reduced consumption of T3 and T4 in the culture medium of HTR-8/Svneo cells treated with DEHP at 400 μM. DEHP treatment did not affect T3 uptake and the expression of monocarboxylate transporters 8 (MCT8) and organic anion transporters 1C1 (OATP1C1). However, DEHP significantly inhibited transthyretin (TTR) internalization, down-regulated TTR, deiodinase 2 (DIO2), and thyroid hormone receptors mRNA expression and protein levels, and up-regulated deiodinase 3 (DIO3) protein levels in a dose-dependent manner. These results indicate that DEHP acts on placental trophoblast cells, inhibits its TTR internalization, down-regulates TTR expression and affects the expression of DIO2, DIO3, and thyroid hormone receptor. These may be the mechanisms by which PAEs affects THs transport through placental.
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%