Merck
  • Microinjection of a growth factor cocktail affects activated microglia in the neocortex of adult rats.
Microinjection of a growth factor cocktail affects activated microglia in the neocortex of adult rats.
Neural regeneration research (2020-03-27)
Ruo-Xu Liu, Jie Ma, Ning Guo, Shao-Jun Liu
ABSTRACT
Microglia, as the resident immune cells in the central nervous system, play important roles in regulating neuronal processes, such as neural excitability, synaptic activity, and apoptotic cell clearance. Growth factors can activate multiple signaling pathways in central nervous system microglia and can regulate their immune effects, but whether growth factors can affect the morphological characteristics and ultrastructure of microglia has not been reported. After microinjecting 300 nL of a growth factor cocktail, including 10 μg/mL epidermal growth factor, 10 μg/mL basic fibroblast growth factor, 10 μg/mL hepatocyte growth factor and 10 μg/mL insulin-like growth factor into adult rat cortex, we found that the number of IBA1-positive microglia around the injection area increased significantly, indicating local activation of microglia. All CD68-positive labeling co-localized with IBA1 in microglia. Cell bodies and protrusions of CD68-positive cells were strongly attached to or were engulfing neurons. Characteristic huge phagosomes were observed in activated phagocytes by electron microscopy. The phagosomes generally included non-degraded neuronal protrusions and mitochondria, yet they contained no myelin membrane or remnants, which might indicate selective phagocytosis by the phagocytes. The remnant myelin sheath after phagocytosis still had regenerative ability and formed "myelin-like" structures around phagocytes. These results show that microinjection of a growth factor cocktail into the cerebral cortex of rodents can locally activate microglia and induce selective phagocytosis of neural structures by phagocytes. The study was approved by the Institute of Laboratory Animal Science, Beijing Institute of Basic Medical Sciences (approval No. IACUC-AMMS-2014-501) on June 30, 2014.
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
IGF-I Protein, Human Recombinant Animal Free, The Insulin-like growth factor I protein(or IGF-1 protein), also known as somatomedin C, is the dominant effector of growth hormone & is structurally homologous to proinsulin. Manufactured using all non-animal reagents.
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture