Merck
  • DNA Methylation Changes are Associated with the Programming of White Adipose Tissue Browning Features by Resveratrol and Nicotinamide Riboside Neonatal Supplementations in Mice.
DNA Methylation Changes are Associated with the Programming of White Adipose Tissue Browning Features by Resveratrol and Nicotinamide Riboside Neonatal Supplementations in Mice.
Nutrients (2020-02-16)
Alba Serrano, Madhu Asnani-Kishnani, Charlene Couturier, Julien Astier, Andreu Palou, Jean-François Landrier, Joan Ribot, M Luisa Bonet
ABSTRACT
Neonatal supplementation with resveratrol (RSV) or nicotinamide riboside (NR) programs in male mice brown adipocyte-like features in white adipose tissue (WAT browning) together with improved metabolism in adulthood. We tested the involvement in this programming of long-term epigenetic changes in two browning-related genes that are overexpressed in WAT of supplemented mice, Slc27a1 and Prdm16. Suckling mice received orally the vehicle, RSV or NR from postnatal days 2-to-20. After weaning (d21) onto a chow diet, male mice were habituated to a normal-fat diet (NFD) starting d75, and split on d90 into continuation on the NFD or switching to a high-fat diet (HFD) until euthanization on d164. CpG methylation by bisulfite-sequencing was analyzed on inguinal WAT. Both treatments modified methylation marks in Slc27a1 and Prdm16 and the HFD-dependent dynamics of these marks in the adult WAT, with distinct and common effects. The treatments also affected gene expression of de novo DNA methylases in WAT of young animals (euthanized at d35 in independent experiments). Studies in 3T3-L1 adipocytes indicated the direct effects of RSV and NR on the DNA methylation machinery and favoring browning features. The results support epigenetic effects being involved in WAT programming by neonatal RSV or NR supplementation in male mice.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
JumpStart REDTaq® ReadyMix Reaction Mix, for PCR
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)