Merck
  • Protein tyrosine phosphatase 4A3 (PTP4A3/PRL-3) drives migration and progression of T-cell acute lymphoblastic leukemia in vitro and in vivo.
Protein tyrosine phosphatase 4A3 (PTP4A3/PRL-3) drives migration and progression of T-cell acute lymphoblastic leukemia in vitro and in vivo.
Oncogenesis (2020-02-01)
M Wei, M G Haney, D R Rivas, J S Blackburn
ABSTRACT
T-cell acute lymphoblastic leukemia (T-ALL) is an aggressive blood cancer. There are no immunotherapies and few molecularly targeted therapeutics available for treatment of this malignancy. The identification and characterization of genes and pathways that drive T-ALL progression are critical for the development of new therapies for T-ALL. Here, we determined that the protein tyrosine phosphatase 4A3 (PTP4A3 or PRL-3) plays a critical role in T-ALL initiation and progression by promoting leukemia cell migration. PRL-3 is highly expressed in patient T-ALL samples at both the mRNA and protein levels compared to normal lymphocytes. Knock-down of PRL-3 expression using short-hairpin RNA (shRNA) in human T-ALL cell lines significantly impeded T-ALL cell migration capacity in vitro and reduced their ability to engraft and proliferate in vivo in xenograft mouse models. Additionally, PRL-3 overexpression in a Myc-induced zebrafish T-ALL model significantly accelerated disease onset and shortened the time needed for cells to enter blood circulation. Reverse-phase protein array (RPPA) and gene set enrichment analysis (GSEA) revealed that the SRC signaling pathway is affected by PRL-3. Immunoblot analyses validated that manipulation of PRL-3 expression in T-ALL cells affected the SRC signaling pathway, which is directly involved in cell migration, although Src was not a direct substrate of PRL-3. More importantly, T-ALL cell growth and migration were inhibited by small molecule inhibition of PRL-3, suggesting that PRL-3 has potential as a therapeutic target in T-ALL. Taken together, our study identifies PRL-3 as an oncogenic driver in T-ALL both in vitro and in vivo and provides a strong rationale for targeted therapies that interfere with PRL-3 function.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
SU 6656, ≥98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes