Merck
  • Exercise-induced α-ketoglutaric acid stimulates muscle hypertrophy and fat loss through OXGR1-dependent adrenal activation.
Exercise-induced α-ketoglutaric acid stimulates muscle hypertrophy and fat loss through OXGR1-dependent adrenal activation.
The EMBO journal (2020-02-28)
Yexian Yuan, Pingwen Xu, Qingyan Jiang, Xingcai Cai, Tao Wang, Wentong Peng, Jiajie Sun, Canjun Zhu, Cha Zhang, Dong Yue, Zhihui He, Jinping Yang, Yuxian Zeng, Man Du, Fenglin Zhang, Lucas Ibrahimi, Sarah Schaul, Yuwei Jiang, Jiqiu Wang, Jia Sun, Qiaoping Wang, Liming Liu, Songbo Wang, Lina Wang, Xiaotong Zhu, Ping Gao, Qianyun Xi, Cong Yin, Fan Li, Guli Xu, Yongliang Zhang, Gang Shu
ABSTRACT
Beneficial effects of resistance exercise on metabolic health and particularly muscle hypertrophy and fat loss are well established, but the underlying chemical and physiological mechanisms are not fully understood. Here, we identified a myometabolite-mediated metabolic pathway that is essential for the beneficial metabolic effects of resistance exercise in mice. We showed that substantial accumulation of the tricarboxylic acid cycle intermediate α-ketoglutaric acid (AKG) is a metabolic signature of resistance exercise performance. Interestingly, human plasma AKG level is also negatively correlated with BMI. Pharmacological elevation of circulating AKG induces muscle hypertrophy, brown adipose tissue (BAT) thermogenesis, and white adipose tissue (WAT) lipolysis in vivo. We further found that AKG stimulates the adrenal release of adrenaline through 2-oxoglutarate receptor 1 (OXGR1) expressed in adrenal glands. Finally, by using both loss-of-function and gain-of-function mouse models, we showed that OXGR1 is essential for AKG-mediated exercise-induced beneficial metabolic effects. These findings reveal an unappreciated mechanism for the salutary effects of resistance exercise, using AKG as a systemically derived molecule for adrenal stimulation of muscle hypertrophy and fat loss.
Sigma-Aldrich
MISSION® esiRNA, targeting human OXGR1
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
SR 59230A, ≥98% (HPLC), powder
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source