Merck
  • The cholesterol metabolite 27-hydroxycholesterol promotes atherosclerosis via proinflammatory processes mediated by estrogen receptor alpha.
The cholesterol metabolite 27-hydroxycholesterol promotes atherosclerosis via proinflammatory processes mediated by estrogen receptor alpha.
Cell metabolism (2014-06-24)
Michihisa Umetani, Pritam Ghosh, Tomonori Ishikawa, Junko Umetani, Mohamed Ahmed, Chieko Mineo, Philip W Shaul
ABSTRACT
Oxysterols are cholesterol metabolites that serve multiple functions in lipid metabolism, including as liver X receptor (LXR) ligands. 27-hydroxycholesterol (27HC) is an abundant oxysterol metabolized by CYP7B1. How 27HC impacts vascular health is unknown. We show that elevations in 27HC via cyp7b1 deletion promote atherosclerosis in apoe(-/-) mice without altering lipid status; furthermore, estrogen-related atheroprotection is attenuated. In wild-type mice, leukocyte-endothelial cell adhesion is increased by 27HC via estrogen receptor (ER)-dependent processes. In monocytes/macrophages, 27HC upregulates proinflammatory genes and increases adhesion via ERα. In endothelial cells, 27HC is also proadhesive via ERα, and in contrast to estrogen, which blunts NF-κB activation, 27HC stimulates NF-κB activation via Erk1,2 and JNK-dependent IκBα degradation. Whereas 27HC administration to apoe(-/-) mice increases atherosclerosis, apoe(-/-);erα(-/-) are unaffected. Thus, 27HC promotes atherosclerosis via proinflammatory processes mediated by ERα, and it attenuates estrogen-related atheroprotection. Strategies to lower 27HC may complement approaches targeting cholesterol to prevent vascular disease.
Product Description
Avanti
27-hydroxycholesterol, Avanti Research - A Croda Brand