Merck
  • 23,24-Dihydrocucurbitacin B promotes lipid clearance by dual transcriptional regulation of LDLR and PCSK9.
23,24-Dihydrocucurbitacin B promotes lipid clearance by dual transcriptional regulation of LDLR and PCSK9.
Acta pharmacologica Sinica (2019-07-31)
Hui-Hui Li, Jun Li, Xian-Jing Zhang, Jiao-Meng Li, Cong Xi, Wen-Qiong Wang, You-Li Lu, Li-Jiang Xuan
ABSTRACT
23,24-Dihydrocucurbitacin B (designated as C95 in this article) is a cucurbitane triterpenoid that has been shown to possess a variety of pharmacological activities, such as anti-inflammatory and anti-HIV-1 activities etc. In this study, we investigated the effects of 23,24-dihydrocucurbitacin B on lipid regulation. We showed that 23,24-dihydrocucurbitacin B (1-5 μM) dose-dependently promoted DiI-LDL uptake in HepG2 cells by upregulating low-density lipoprotein receptor (LDLR) protein. In HepG2 cells, 23,24-dihydrocucurbitacin B (1-10 μM) dose-dependently enhanced LDLR promoter activity by elevating the mature form of SREBP2 (sterol regulatory element binding protein 2) protein levels on one hand, and inhibited PCSK9 (proprotein convertase subtilisin/kexin type 9) promoter activity by attenuating HNF1α (hepatocyte nuclear factor-1α) protein levels in nuclei on the other hand. Consequently, the expression of LDLR protein markedly increased, whereas the PCSK9-mediated LDLR protein degradation decreased. In a high-cholesterol LVG golden Syrian Hamster model, administration of 23,24-dihydrocucurbitacin B (30 mg · kg-1⋅ d-1, intragastric, for 3 weeks) significantly decreased the serum LDL-cholesterol (LDL-C) levels. PCSK9 protein levels in the serum and liver tissues were significantly decreased, whereas LDLR protein levels in liver tissues were significantly increased in the treated animals as compared with the control animals. In conclusion, our study demonstrates for the first time that 23,24-dihydrocucurbitacin B exhibits dual transcriptional regulation of LDLR and PCSK9 in HepG2 cells by increasing SREBP2 protein levels and decreasing HNF1α protein levels in the nuclei. These results propose a new strategy to simultaneously manage LDLR and PCSK9 protein expression and provide a promising lead compound for drug development.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.