Merck
  • Evaluation of PAX8 expression promotes the proliferation of stomach Cancer cells.
Evaluation of PAX8 expression promotes the proliferation of stomach Cancer cells.
BMC molecular and cell biology (2019-12-29)
Liang-Yu Bie, Ning Li, Wen-Ying Deng, Xiao-Yu Lu, Ping Guo, Su-Xia Luo
ABSTRACT
PAX8 was not only a mitotic factor, but identified as a transcription factor involved in the prognosis of human tumor patients. Elucidating the function of PAX8 on the pathology of stomach cancer was meaningful. PAX8 was found to be upregulated in primary stomach cancer tissue and the TCGA stomach cancer dataset. Interestingly, SOX13 and PAX8 showed consistent expression patterns, and the combined high PAX8 and SOX18 expression induced a worse prognosis of stomach cancer patients. SOX13 was further identified as a transcription factor of PAX8, and further affect Aurora B and Cyclin B1 expression, two cell cycle related factors of the downstream of PAX8, including. Furthermore, PAX8 depletion inducted G1-phase arrest and the decrease of EdU incorporation, cell viability and colony formation can be rescued by SOX13 overexpression. SOX13 participated in the elevated expression of PAX8, which promote the proliferation of stomach cancer cells. Therefore, SOX13 mediated PAX8 expression was recognized as a tumor-promoting role in stomach cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human SOX13
Pricing and availability is not currently available.