Merck
  • Perineuronal Net Formation during the Critical Period for Neuronal Maturation in the Hypothalamic Arcuate Nucleus.
Perineuronal Net Formation during the Critical Period for Neuronal Maturation in the Hypothalamic Arcuate Nucleus.
Nature metabolism (2019-06-28)
Zaman Mirzadeh, Kimberly M Alonge, Elaine Cabrales, Vicente Herranz-Pérez, Jarrad M Scarlett, Jenny M Brown, Rim Hassouna, Miles E Matsen, Hong T Nguyen, Jose Manuel Garcia-Verdugo, Lori M Zeltser, Michael W Schwartz
ABSTRACT
In leptin-deficient ob/ob mice, obesity and diabetes are associated with abnormal development of neurocircuits in the hypothalamic arcuate nucleus (ARC)1, a critical brain area for energy and glucose homeostasis2,3. As this developmental defect can be remedied by systemic leptin administration, but only if given before postnatal day 28, a critical period (CP) for leptin-dependent development of ARC neurocircuits has been proposed4. In other brain areas, CP closure coincides with the appearance of perineuronal nets (PNNs), extracellular matrix specializations that restrict the plasticity of neurons that they enmesh5. Here we report that in humans as well as rodents, subsets of neurons in the mediobasal aspect of the ARC are enmeshed by PNN-like structures. In mice, these neurons are densely-packed into a continuous ring that encircles the junction of the ARC and median eminence, which facilitates exposure of ARC neurons to the circulation. Most of the enmeshed neurons are both GABAergic and leptin receptor-positive, including a majority of Agrp neurons. Postnatal formation of the PNN-like structures coincides precisely with closure of the CP for Agrp neuron maturation and is dependent on input from circulating leptin, as postnatal ob/ob mice have reduced ARC PNN-like material that is restored by leptin administration during the CP. We conclude that neurons crucial to metabolic homeostasis are enmeshed by PNN-like structures and organized into a densely packed cluster situated circumferentially at the ARC-ME junction, where metabolically-relevant humoral signals are sensed.
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®
Sigma-Aldrich
Anti-DSD-1 Antibody, clone 473HD, clone 473HD, from rat