Merck
  • Lysis of dysplastic but not normal oral keratinocytes and tissue-engineered epithelia with conditionally replicating adenoviruses.
Lysis of dysplastic but not normal oral keratinocytes and tissue-engineered epithelia with conditionally replicating adenoviruses.
Cancer research (2007-08-03)
Kamis Gaballah, Allison Hills, David Curiel, Gunnel Hallden, Paul Harrison, Max Partridge
ABSTRACT
There is no effective medical treatment for oral precancer, and surgery to remove these lesions is imprecise because abnormal mucosa extends beyond the visible lesion. Development of vectors for tumor-selective viral replication has been a significant advance, and viral lysis is well suited to destruction of oral precancerous mucosa. To facilitate evaluation of new treatments, we engineered dysplastic oral epithelium using keratinocytes isolated from dysplastic lesions. We show that these model systems recapitulate the key characteristics of the clinical lesions closely, and that topical delivery of the conditionally replicating adenovirus (CRAd) dl922-947 can lyse tissue-engineered epithelia that show mild, moderate, or severe dysplasia, but normal oral epithelia are very resistant to this treatment. The lytic effect is determined by various factors, including the grade and proliferation index of the dysplastic epithelia. The presence of suprabasal cycling cells, expression of the coxsackie adenovirus receptor (CAR), the transcription cofactor p300, and other aberrations that affect the regulation of the cell cycle or apoptosis and promote viral replication may also be important. The ability of dl922-947 to destroy engineered oral dysplasia was significantly greater than that observed using wild-type adenovirus, d/1520, or viruses modified to bypass cell entry dependent on the presence of CAR. Evidence of infection in clinical dysplastic lesions was also shown ex vivo using tissue explants. We conclude that dl922-947 may provide an efficient molecular cytotoxic to dissolve oral dysplastic lesions.
Sigma-Aldrich
Anti-Laminin-5 (γ2 chain) Antibody, clone D4B5, clone D4B5, Chemicon®, from mouse