Merck
  • Structural basis for adhesion G protein-coupled receptor Gpr126 function.
Structural basis for adhesion G protein-coupled receptor Gpr126 function.
Nature communications (2020-01-12)
Katherine Leon, Rebecca L Cunningham, Joshua A Riback, Ezra Feldman, Jingxian Li, Tobin R Sosnick, Minglei Zhao, Kelly R Monk, Demet Araç
ABSTRACT
Many drugs target the extracellular regions (ECRs) of cell-surface receptors. The large and alternatively-spliced ECRs of adhesion G protein-coupled receptors (aGPCRs) have key functions in diverse biological processes including neurodevelopment, embryogenesis, and tumorigenesis. However, their structures and mechanisms of action remain unclear, hampering drug development. The aGPCR Gpr126/Adgrg6 regulates Schwann cell myelination, ear canal formation, and heart development; and GPR126 mutations cause myelination defects in human. Here, we determine the structure of the complete zebrafish Gpr126 ECR and reveal five domains including a previously unknown domain. Strikingly, the Gpr126 ECR adopts a closed conformation that is stabilized by an alternatively spliced linker and a conserved calcium-binding site. Alternative splicing regulates ECR conformation and receptor signaling, while mutagenesis of the calcium-binding site abolishes Gpr126 function in vivo. These results demonstrate that Gpr126 ECR utilizes a multi-faceted dynamic approach to regulate receptor function and provide relevant insights for ECR-targeted drug design.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Seleno-L-methionine, ≥98% (TLC), powder
Whatman® qualitative filter paper, Grade 1, circles, diam. 110 mm, pack of 100
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep