Merck
  • TIMP1 mRNA in tumor-educated platelets is diagnostic biomarker for colorectal cancer.
TIMP1 mRNA in tumor-educated platelets is diagnostic biomarker for colorectal cancer.
Aging (2019-10-23)
Liu Yang, Qian Jiang, Dong-Zheng Li, Xin Zhou, Dong-Sheng Yu, Jian Zhong
ABSTRACT
Platelets have been shown to promote the growth of tumors, including colorectal cancer. The RNA profile of tumor-educated platelets has the possibility for cancer diagnosis. We used RNA sequencing to identified the gene expression signature in platelets from colorectal cancer patients and healthy volunteers. We then verified the selected biomarkers from the RNA sequencing in a two-step case-control study using quantitative reverse-transcription polymerase chain reaction. We found that TIMP1 mRNA levels are higher in platelets from colorectal cancer patients than in platelets from healthy volunteers and patients with inflammatory bowel diseases. Additionally, TIMP1 mRNA expressed in platelets from colorectal cancer patients can be carried into colorectal cancer cells, where it promotes tumor growth in vivo and in vitro. These findings show that the TIMP1 mRNA in platelets is a potential independent diagnostic biomarker for colorectal cancer, and that platelets can carry RNAs into colorectal cancer cells to promote colorectal cancer development.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-TIMP1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Human TIMP-1 ELISA Kit, for cell and tissue lysates