Merck
Distribution of bioactive compounds in pearled fractions of tritordeum.
Food chemistry (2019-08-05)
Debora Giordano, Amedeo Reyneri, Monica Locatelli, Jean Daniel Coïsson, Massimo Blandino
ABSTRACT
Hexaploid tritordeum is the amphidiploid cereal derived from the cross between wild barley and durum wheat. The present study compares two cultivars of tritordeum with other cereals grown in the same experimental area to weigh up its potential use as ingredient for health-valued foods. Tritordeum shows 2.5-fold higher concentration of lutein than common wheat and barley, and 1.2-fold higher than durum wheat, while the concentration of β-glucans is 5 folds lower than the one observed for barley. Based on the distribution of bioactive compounds in pearled fractions, the use of whole-grain flours seems the best way to exploit the antioxidant potential of tritordeum. Nevertheless, the internal layers of the kernel of this cereal are characterized on average by high concentrations of antioxidants (32.0 mg/kg and 518 mg/kg soluble and cell wall-bound phenolic acids, respectively), making tritordeum interesting also for the production of refined flours rich in bioactive compounds.
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%