Merck
  • Ionizing radiation increases the endothelial permeability and the transendothelial migration of tumor cells through ADAM10-activation and subsequent degradation of VE-cadherin.
Ionizing radiation increases the endothelial permeability and the transendothelial migration of tumor cells through ADAM10-activation and subsequent degradation of VE-cadherin.
BMC cancer (2019-10-18)
Pascaline Nguemgo Kouam, Günther A Rezniczek, Irenäus A Adamietz, Helmut Bühler
ABSTRACT
We analyzed the changes in permeability of endothelial cell layers after photon irradiation, with a focus on the metalloproteases ADAM10 and ADAM17, and on VE-cadherin, components crucial for the integrity of endothelial intercellular junctions, and their roles in the transmigration of cancer cells through endothelial cell monolayers. Primary HUVEC were irradiated with 2 or 4 Gy photons at a dose rate of 5 Gy/min. The permeability of an irradiated endothelial monolayer for macromolecules and tumor cells was analyzed in the presence or absence of the ADAM10/17 inhibitors GI254023X and GW280264X. Expression of ADAM10, ADAM17 and VE-Cadherin in endothelial cells was quantified by immunoblotting and qRT. VE-Cadherin was additionally analyzed by immunofluorescence microscopy and ELISA. Ionizing radiation increased the permeability of endothelial monolayers and the transendothelial migration of tumor cells. This was effectively blocked by a selective inhibition (GI254023X) of ADAM10. Irradiation increased both, the expression and activity of ADAM10, which led to increased degradation of VE-cadherin, but also led to higher rates of VE-cadherin internalization. Increased degradation of VE-cadherin was also observed when endothelial monolayers were exposed to tumor-cell conditioned medium, similar to when exposed to recombinant VEGF. Our results suggest a mechanism of irradiation-induced increased permeability and transendothelial migration of tumor cells based on the activation of ADAM10 and the subsequent change of endothelial permeability through the degradation and internalization of VE-cadherin.
Sigma-Aldrich
Vascular Endothelial Growth Factor from mouse, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.
Sigma-Aldrich
N-(3-Aminopropyl)methacrylamide hydrochloride, contains ≤1,000 ppm MEHQ as stabilizer, 98% (HPLC)
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
GI254023X, ≥98% (HPLC)