Merck
Linking YAP to Müller Glia Quiescence Exit in the Degenerative Retina.
Cell reports (2019-05-09)
Annaïg Hamon, Diana García-García, Divya Ail, Juliette Bitard, Albert Chesneau, Deniz Dalkara, Morgane Locker, Jérôme E Roger, Muriel Perron
ABSTRACT
Contrasting with fish or amphibian, retinal regeneration from Müller glia is largely limited in mammals. In our quest toward the identification of molecular cues that may boost their stemness potential, we investigated the involvement of the Hippo pathway effector YAP (Yes-associated protein), which is upregulated in Müller cells following retinal injury. Conditional Yap deletion in mouse Müller cells prevents cell-cycle gene upregulation that normally accompanies reactive gliosis upon photoreceptor cell death. We further show that, in Xenopus, a species endowed with efficient regenerative capacity, YAP is required for their injury-dependent proliferative response. In the mouse retina, where Müller cells do not spontaneously proliferate, YAP overactivation is sufficient to induce their reprogramming into highly proliferative cells. Overall, we unravel a pivotal role for YAP in tuning Müller cell proliferative response to injury and highlight a YAP-EGFR (epidermal growth factor receptor) axis by which Müller cells exit their quiescence state, a critical step toward regeneration.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone RET-P1, clone RET-P1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone 4D2, clone 4D2, from mouse
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Calretinin Antibody, clone 6B8.2, clone 6B8.2, Chemicon®, from mouse