Merck
  • Organic contamination of Italian and Tunisian culinary herbs and spices.
Organic contamination of Italian and Tunisian culinary herbs and spices.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-02-19)
Giuseppa Di Bella, Angela Giorgia Potortì, Asma Ben Tekaya, Asma Beltifa, Hedi Ben Mansour, Emanuele Sajia, Giovanni Bartolomeo, Clara Naccari, Giacomo Dugo, Vincenzo Lo Turco
ABSTRACT
Contaminants belonging to various classes, as polychlorobiphenyls, polycyclic aromatic hydrocarbons, organochlorine pesticides, organophosphorous pesticides, pyrethroid insecticides, fungicides, herbicides, synergists, carbamates, acaricides and insect growth regulators were simultaneously analysed by GC-MS/MS in 118 Italian and Tunisian culinary herbs and spices. The results obtained in Italian samples shown that laurel samples were the most contaminated with the presence of 15 residues on 140, whereas in rosemary (max value of 35 ng/g for cis-chlorfenyvinphos) and oregano (max value of 118.16 ng/g for ethion) some occasional residues can be observed, but always lower than the maximum residue levels; all the others samples shown no contamination. Among Tunisian samples, only rosemary contains a notably high content of pollutants exceeded the EU maximum residue limits (i.e., alachlor and phosalone with level of 359.2 and 43.3 ng/g, respectively), while oregano was determined to be free of contaminant residues. Considering the comparison among the different organic pollutants in Italian and Tunisian spices and herbs evaluated and the differences observed in this study a harmonization of regulation on contaminant residues in herbs and spices for human consumption should be needed, considering their increased use in diet and cooking.