Merck
  • Design, Synthesis, Docking Studies and Monoamine Oxidase Inhibition of a Small Library of 1-acetyl- and 1-thiocarbamoyl-3,5-diphenyl-4,5-dihydro-(1H)-pyrazoles.
Design, Synthesis, Docking Studies and Monoamine Oxidase Inhibition of a Small Library of 1-acetyl- and 1-thiocarbamoyl-3,5-diphenyl-4,5-dihydro-(1H)-pyrazoles.
Molecules (Basel, Switzerland) (2019-02-01)
Paolo Guglielmi, Simone Carradori, Giulio Poli, Daniela Secci, Roberto Cirilli, Giulia Rotondi, Paola Chimenti, Anél Petzer, Jacobus P Petzer
ABSTRACT
New N-acetyl/N-thiocarbamoylpyrazoline derivatives were designed and synthesized in high yields to assess their inhibitory activity and selectivity against human monoamine oxidase A and B. The most important chiral compounds were separated into their single enantiomers and tested. The impact of the substituents at N1, C3 and C5 positions as well the influence of the configuration of the C5 on the biological activity were analyzed. Bulky aromatic groups at C5 were not tolerated. p-Prenyloxyaryl moiety at C3 oriented the selectivity toward the B isoform. The results were also corroborated by molecular modelling studies providing new suggestions for the synthesis of privileged structures to serve as lead compounds for the treatment of mood disorders and neurodegenerative diseases.