Merck
  • Protease-Activatable Adeno-Associated Virus Vector for Gene Delivery to Damaged Heart Tissue.
Protease-Activatable Adeno-Associated Virus Vector for Gene Delivery to Damaged Heart Tissue.
Molecular therapy : the journal of the American Society of Gene Therapy (2019-02-18)
Caitlin M Guenther, Mitchell J Brun, Antonette D Bennett, Michelle L Ho, Weitong Chen, Banghe Zhu, Michael Lam, Momona Yamagami, Sunkuk Kwon, Nilakshee Bhattacharya, Duncan Sousa, Annicka C Evans, Julie Voss, Eva M Sevick-Muraca, Mavis Agbandje-McKenna, Junghae Suh
ABSTRACT
Adeno-associated virus (AAV) has emerged as a promising gene delivery vector because of its non-pathogenicity, simple structure and genome, and low immunogenicity compared to other viruses. However, its adoption as a safe and effective delivery vector for certain diseases relies on altering its tropism to deliver transgenes to desired cell populations. To this end, we have developed a protease-activatable AAV vector, named provector, that responds to elevated extracellular protease activity commonly found in diseased tissue microenvironments. The AAV9-based provector is initially inactive, but then it can be switched on by matrix metalloproteinases (MMP)-2 and -9. Cryo-electron microscopy and image reconstruction reveal that the provector capsid is structurally similar to that of AAV9, with a flexible peptide insertion at the top of the 3-fold protrusions. In an in vivo model of myocardial infarction (MI), the provector is able to deliver transgenes site specifically to high-MMP-activity regions of the damaged heart, with concomitant decreased delivery to many off-target organs, including the liver. The AAV provector may be useful in the future for enhanced delivery of transgenes to sites of cardiac damage.
Sigma-Aldrich
Anti-RANTES antibody produced in goat, affinity isolated antibody, lyophilized powder