- Microwave-Assisted Hydrothermal Synthesis of [Al(OH)(1,4-NDC)] Membranes with Superior Separation Performances.
Microwave-Assisted Hydrothermal Synthesis of [Al(OH)(1,4-NDC)] Membranes with Superior Separation Performances.
Chemistry, an Asian journal (2019-05-09)
Yi Liu, Akihiro Hori, Shinpei Kusaka, Nobuhiko Hosono, Mingrun Li, Ang Guo, Dongying Du, Yanshuo Li, Weishen Yang, Yunsheng Ma, Ryotaro Matsuda
PMID31066192
ABSTRACT
In this study, we report a facile ligand-assisted in situ hydrothermal approach for preparation of compact [Al(OH)(1,4-NDC)] (1,4-NDC=1,4-naphthalenedicarboxylate) MOF membranes on porous γ-Al2 O3 substrates, which also served as the Al3+ source of MOF membranes. Simultaneously, it was observed that the heating mode exerted significant influence on the final microstructure and separation performance of [Al(OH)(1,4-NDC)] membranes. Compared with the conventional hydrothermal method, the employment of microwave heating led to the formation of [Al(OH)(1,4-NDC)] membranes composed of closely packed nanorods with superior H2 /CH4 selectivity.