Merck
  • Increased Cholesterol Biosynthesis Is a Key Characteristic of Breast Cancer Stem Cells Influencing Patient Outcome.
Increased Cholesterol Biosynthesis Is a Key Characteristic of Breast Cancer Stem Cells Influencing Patient Outcome.
Cell reports (2019-06-27)
Sidse Ehmsen, Martin H Pedersen, Guisong Wang, Mikkel G Terp, Amina Arslanagic, Brian L Hood, Thomas P Conrads, Rikke Leth-Larsen, Henrik J Ditzel
ABSTRACT
Tumor eradication may be greatly improved by targeting cancer stem cells (CSCs), as they exhibit resistance to conventional therapy. To gain insight into the unique biology of CSCs, we developed patient-derived xenograft tumors (PDXs) from ER- breast cancers from which we isolated mammospheres that are enriched for CSCs. Comparative global proteomic analysis was performed on patient tumor tissues and corresponding PDXs and mammospheres. Mammospheres exhibited increased expression of proteins associated with de novo cholesterol synthesis. The clinical relevance of increased cholesterol biosynthesis was verified in a large breast cancer cohort showing correlation with shorter relapse-free survival. RNAi and chemical inhibition of the cholesterol biosynthesis pathway reduced mammosphere formation, which could be rescued by a downstream metabolite. Our findings identify the cholesterol biosynthesis pathway as central for CSC propagation and a potential therapeutic target, as well as providing a mechanistic explanation for the therapeutic benefit of statins in breast cancer.
Sigma-Aldrich
Simvastatin, A lipophilic 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that blocks Ras function through inhibition of farnesylation.
Sigma-Aldrich
Anti-FDFT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CYB5R3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution